accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COCH3NHH3C

CAS RN 2386-25-6

IUPAC Name: 1-(2,4-dimethyl-1H-pyrrol-3-yl)ethan-1-one
Synonyms: 1-(2,4-dimethyl-1H-pyrrol-3-yl)ethanone
Molecular Weight (g/mol): 137.18
Molecular Formula: C8H11NO
InChi Key: VGZCKCJMYREIKA-UHFFFAOYSA-N
SMILES: CC(=O)C1=C(C)NC=C1C
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results