CAS RN 24388-68-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Ethylene Terephthalate Cyclic Dimer, TRC
CAS: 24388-68-9 Molecular Formula: C20 H16 O8 Molecular Weight (g/mol): 384.3362 Synonym: 3,6,13,16-Tetraoxatricyclo[16.2.2.28,11]tetracosa-8,10,18,20,21,23-hexaene-2,7,12,17-tetrone,Terephthalic Acid, Bimol. Cyclic Ethylene Ester,Ethylene Glycol, Bimol. Cyclic Terephthalate SMILES: O=C1OCCOC(=O)c2ccc(cc2)C(=O)OCCOC(=O)c3ccc1cc3