CAS RN 2444-89-5
IUPAC Name:
1-chloro-4-(4-chlorophenoxy)benzene
Synonyms:
4,4'-dichlorodiphenyl ether
Molecular Weight (g/mol):
239.1
Molecular Formula:
C12H8Cl2O
InChi Key:
URUJZHZLCCIILC-UHFFFAOYSA-N
SMILES:
ClC1=CC=C(OC2=CC=C(Cl)C=C2)C=C1
1
–
1
of
1
results
1
4,4'-Dichlorodiphenyl Ether, TRC
CAS: 2444-89-5 Molecular Formula: C12H8Cl2O Molecular Weight (g/mol): 239.1 Synonym: 1,1'-Oxybis[4-chlorobenzene],Bis(4-chlorophenyl) Ether,Bis(p-chlorophenyl) Ether,CDE-15,NSC 38001,PCDE 15,p,p'-Dichlorodiphenyl Oxide SMILES: Clc1ccc(Oc2ccc(Cl)cc2)cc1