missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Dichlorodiphenyl Ether, TRC

Product Code. 30378548
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30378548 100 mg 100mg
30375506 500 mg -
30369129 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30378548 Supplier Toronto Research Chemicals Supplier No. TRCD438540100MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2444-89-5
Molecular Formula C12H8Cl2O
Molecular Weight (g/mol) 239.1
Synonym 1,1'-Oxybis[4-chlorobenzene], Bis(4-chlorophenyl) Ether, Bis(p-chlorophenyl) Ether, CDE-15, NSC 38001, PCDE 15, p,p'-Dichlorodiphenyl Oxide
SMILES Clc1ccc(Oc2ccc(Cl)cc2)cc1

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C12H8Cl2O/c13-9-1-5-11(6-2-9)15-12-7-3-10(14)4-8-12/h1-8H
Formula Weight 237.9952
Chemical Name or Material 4,4'-Dichlorodiphenyl Ether
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.