accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 2454-37-7

H3COHH2N

CAS RN 2454-37-7

IUPAC Name: 1-(3-aminophenyl)ethan-1-ol
Synonyms: 1-(3-aminophenyl)ethanol
Molecular Weight (g/mol): 137.18
Molecular Formula: C8H11NO
InChi Key: QPKNDHZQPGMLCJ-UHFFFAOYNA-N
SMILES: CC(O)C1=CC=CC(N)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results