accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 24621-70-3

HOHN

CAS RN 24621-70-3

IUPAC Name: (1H-indol-2-yl)methanol
Synonyms: 1H-indol-2-ylmethanol
Molecular Weight (g/mol): 147.18
Molecular Formula: C9H9NO
InChi Key: XEEANGGQJOWRTG-UHFFFAOYSA-N
SMILES: OCC1=CC2=CC=CC=C2N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results