accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 2476-29-1

IUPAC Name: 1-(4-amino-2-hydroxyphenyl)ethan-1-one
Synonyms: 1-(4-amino-2-hydroxyphenyl)ethanone
Molecular Weight (g/mol): 151.17
Molecular Formula: C8H9NO2
InChi Key: QQZFVONVJPXCSQ-UHFFFAOYSA-N
SMILES: CC(=O)C1=C(O)C=C(N)C=C1
Quantity 
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results