accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OHOBr

CAS RN 2491-37-4

IUPAC Name: 2-bromo-1-(3-hydroxyphenyl)ethan-1-one
Synonyms: 2-bromo-1-(3-hydroxyphenyl)ethanone
Molecular Weight (g/mol): 215.05
Molecular Formula: C8H7BrO2
InChi Key: IEPSGFQQGKPTPM-UHFFFAOYSA-N
SMILES: OC1=CC=CC(=C1)C(=O)CBr
Quantity 
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results