accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 24947-68-0

H3N(S)(S)HO

CAS RN 24947-68-0

IUPAC Name: [(1S,2S)-2-hydroxycyclohexyl]methanaminium
Synonyms: [(1S,2S)-2-hydroxycyclohexyl]methanaminium
Molecular Weight (g/mol): 130.21
Molecular Formula: C7H16NO
InChi Key: JGKFBZBVCAWDFD-BQBZGAKWSA-O
SMILES: [NH3+]C[C@@H]1CCCC[C@@H]1O
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results