accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 2495-37-6

H3CCH2OO

CAS RN 2495-37-6

IUPAC Name: benzyl 2-methylprop-2-enoate
Synonyms: benzyl methacrylate
Molecular Weight (g/mol): 176.22
Molecular Formula: C11H12O2
InChi Key: AOJOEFVRHOZDFN-UHFFFAOYSA-N
SMILES: CC(=C)C(=O)OCC1=CC=CC=C1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results