accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 24965-90-0

CH3HO

CAS RN 24965-90-0

IUPAC Name: 3-methylcyclohexan-1-ol
Synonyms: 3-methylcyclohexanol
Molecular Weight (g/mol): 114.19
Molecular Formula: C7H14O
InChi Key: HTSABYAWKQAHBT-UHFFFAOYNA-N
SMILES: CC1CCCC(O)C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results