accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COCl

CAS RN 25017-08-7

IUPAC Name: 1-(4-chlorophenyl)pentan-1-one
Synonyms: 1-(4-chlorophenyl)pentan-1-one
Molecular Weight (g/mol): 196.67
Molecular Formula: C11H13ClO
InChi Key: XMUGWCSIQUJOFA-UHFFFAOYSA-N
SMILES: CCCCC(=O)C1=CC=C(Cl)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results