accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CNNHSO

CAS RN 2503-66-4

IUPAC Name: 5-(2-methylphenyl)-2,3-dihydro-1,3,4-oxadiazole-2-thione
Synonyms: 5-(2-methylphenyl)-3H-1,3,4-oxadiazole-2-thione
Molecular Weight (g/mol): 192.24
Molecular Formula: C9H8N2OS
InChi Key: HNFIJWNVPWXSOL-UHFFFAOYSA-N
SMILES: CC1=CC=CC=C1C1=NNC(=S)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results