accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HClNNNH

CAS RN 2508-72-7

IUPAC Name: hydrogen N-benzyl-N-[(4,5-dihydro-1H-imidazol-2-yl)methyl]aniline chloride
Synonyms: antazoline
Molecular Weight (g/mol): 301.82
Molecular Formula: C17H20ClN3
InChi Key: SWKDMSRRIBZZAY-UHFFFAOYSA-N
SMILES: [H+].[Cl-].C(N(CC1=CC=CC=C1)C1=CC=CC=C1)C1=NCCN1
Molecular Weight (g/mol) 
  • (6)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Certifications/Compliance 
  • (2)
  • (1)
Industry Type 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (3)
  • (6)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)