missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Antazoline hydrochloride, 98%

Imidazoline binding site ligand and an antihistamine

Brand:  Alfa Aesar™ J62511.14

Quantity 25g

Product Code. 15454889

Request Bulk or Custom Format

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »

This item is not returnable. View return policy

Description

Description

TRUSTED_SUSTAINABILITY
Specifications

Specifications

2508-72-7
MFCD00058145
14,680
SWKDMSRRIBZZAY-UHFFFAOYSA-N
hydrogen N-benzyl-N-[(4,5-dihydro-1H-imidazol-2-yl)methyl]aniline chloride
17275
98%
C17H20ClN3
25g
antazoline hydrochloride, antazoline hcl, fenazolina, histazine, 2-n-benzylanilinomethyl-2-imidazoline hydrochloride, antazolinium chloratum, antistine hydrochloride, phenazoline hydrochloride, unii-fp8q8f72jh
[H+].[Cl-].C(N(CC1=CC=CC=C1)C1=CC=CC=C1)C1=NCCN1
301.82
301.82
Antazoline hydrochloride
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Promotions

Promotions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.