accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H2OH2OKKHOONNOHOOOOO

CAS RN 25102-12-9

IUPAC Name: dipotassium 2-({2-[(carboxylatomethyl)(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetate dihydrate
Synonyms: dipotassium 2-({2-[(carboxylatomethyl)(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetate dihydrate
Molecular Weight (g/mol): 404.46
Molecular Formula: C10H18K2N2O10
InChi Key: JFROQFOCFOKDKU-UHFFFAOYSA-L
SMILES: O.O.[K+].[K+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O
Molecular Weight (g/mol) 
  • (4)
  • (3)
  • (4)
Quantity 
  • (1)
  • (3)
  • (1)
  • (1)
  • (5)
Percent Purity 
  • (1)
  • (4)
  • (5)
Physical Form 
  • (3)
  • (1)
  • (1)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (1)
  • (3)
  • (2)
  • (4)
  • (4)
  • (3)
  • (4)
  • (1)
  • (3)
  • (1)
  • (1)
  • (5)
  • (1)
  • (4)
  • (5)
  • (3)
  • (1)
  • (1)
  • (3)