missing translation for 'onlineSavingsMsg'
Learn More

Ethylenediaminetetraacetic Acid Dipotassium Salt, Extra Pure, SLR, Fisher Chemical™

Product Code. 10736671 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10736671 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10736671 Supplier Fisher Chemical Supplier No. D/0600/53

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Melting Point 272°C
CAS 25102-12-9
Color White
pH 4.0 to 5.0
Physical Form Powder/Solid
Quantity 500 g
Molecular Formula C10H18K2N2O10
MDL Number 150036
InChI Key JFROQFOCFOKDKU-UHFFFAOYSA-L
SMILES O.O.[K+].[K+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O
IUPAC Name dipotassium 2-({2-[(carboxylatomethyl)(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetate dihydrate
Molecular Weight (g/mol) 404.46
Formula Weight 404.46g/mol
Packaging Plastic powder jar
Show More Show Less
compliance-icons
Product Identifier
  • Ethylenediaminetetraacetic acid, dipotassium salt dihydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.