accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 251096-97-6

IUPAC Name: 1-[3-(trifluoromethyl)benzenesulfonyl]pyrrolidine-2-carboxylic acid
Synonyms: 1-[3-(trifluoromethyl)benzenesulfonyl]pyrrolidine-2-carboxylic acid
Molecular Weight (g/mol): 323.29
Molecular Formula: C12H12F3NO4S
InChi Key: RYNZZKHRKJIGHR-UHFFFAOYNA-N
SMILES: OC(=O)C1CCCN1S(=O)(=O)C1=CC=CC(=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results