missing translation for 'onlineSavingsMsg'
Learn More

1-[3-(Trifluoromethyl)phenylsulfonyl]-L-proline, 96%, Thermo Scientific™

Product Code. 15478665
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15478665 250 mg 250mg
15468665 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15478665 Supplier Thermo Scientific Alfa Aesar Supplier No. H33843.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 251096-97-6
Molecular Formula C12H12F3NO4S
Molecular Weight (g/mol) 323.286
MDL Number MFCD00172809
InChI Key RYNZZKHRKJIGHR-UHFFFAOYSA-N
Synonym 1-3-trifluoromethyl benzenesulfonyl pyrrolidine-2-carboxylic acid, 1-3-trifluoromethyl phenyl sulfonyl-2-pyrrolidinecarboxylic acid, 3-trifluoromethyl phenyl sulfonyl proline, 1-3-trifluoromethyl phenyl sulfonyl pyrrolidine-2-carboxylic acid, 1-3-trifluoromethyl benzenesulphonyl pyrrolidine-2-carboxylic acid, proline,1-3-trifluoromethyl phenyl sulfonyl, 1-3-trifluoromethyl phenyl sulfonylpyrrolidine-2-carboxylic acid, 1-3-trifluoromethyl benzenesulphonylproline
PubChem CID 2777712
IUPAC Name 1-[3-(trifluoromethyl)phenyl]sulfonylpyrrolidine-2-carboxylic acid
SMILES C1CC(N(C1)S(=O)(=O)C2=CC=CC(=C2)C(F)(F)F)C(=O)O

Specifications

Melting Point 101°C to 103°C
Quantity 250 mg
Formula Weight 323.29
Percent Purity 96%
Chemical Name or Material 1-[3-(Trifluoromethyl)phenylsulfonyl]-L-proline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.