accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 25128-35-2

OHOHNOO

CAS RN 25128-35-2

IUPAC Name: 2,3-dioxo-2,3-dihydro-1H-indole-7-carboxylic acid
Synonyms: 2,3-dioxo-1H-indole-7-carboxylic acid
Molecular Weight (g/mol): 191.14
Molecular Formula: C9H5NO4
InChi Key: ROODQCZSWXEDJL-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=CC2=C1NC(=O)C2=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results