CAS RN 2517-43-3
IUPAC Name:
3-methoxybutan-1-ol
Synonyms:
3-methoxybutanol
Molecular Weight (g/mol):
104.15
Molecular Formula:
C5H12O2
InChi Key:
JSGVZVOGOQILFM-UHFFFAOYNA-N
SMILES:
COC(C)CCO
1
–
1
of
1
results
1
3-Methoxy-1-butanol, 99%
CAS: 2517-43-3 Molecular Formula: C5H12O2 Molecular Weight (g/mol): 104.15 MDL Number: MFCD00002931 InChI Key: JSGVZVOGOQILFM-UHFFFAOYNA-N Synonym: 3-methoxy-1-butanol,3-methoxybutanol,1-butanol, 3-methoxy,methoxybutanol,ccris 8976,1-butanol, 3-methoxy-,-,1-butanol, 3-methoxy-, r,3-methoxy-butanol,3-methoxybutane-1-ol,3-methoxybutyl alcohol PubChem CID: 17291 IUPAC Name: 3-methoxybutan-1-ol SMILES: COC(C)CCO