accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 2530-85-0

IUPAC Name: 3-(trimethoxysilyl)propyl 2-methylprop-2-enoate
Synonyms: methacryloxypropyltrimethoxysilane methacryloyl propyltrimethoxysilane
Molecular Weight (g/mol): 248.35
Molecular Formula: C10H20O5Si
InChi Key: XDLMVUHYZWKMMD-UHFFFAOYSA-N
SMILES: CO[Si](CCCOC(=O)C(C)=C)(OC)OC
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (4)
  • (3)
Boiling Point 
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results