Learn More
3-Trimethoxysilyl-Propyl-Methacrylate, Pure, Fisher Chemical
Product Code. 10274160 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
100 g
Packaging:
Amber glass bottle
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10274160 100 g Amber glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10274160 Supplier Fisher Chemical Supplier No. T/3350/48
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
Specifications
CAS 2530-85-0
Boiling Point 190°C
Packaging Amber glass bottle
Molecular Formula C10H20O5Si
MDL Number 8593
Quantity 100 g
Synonym 3-trimethoxysilyl propyl methacrylate, 3-methacryloxypropyltrimethoxysilane, methacryloxypropyltrimethoxysilane, dynasylan memo, silicone a-174, union carbide a-174, mops-m, silane a-174, nuca 174, 2-propenoic acid, 2-methyl-, 3-trimethoxysilyl propyl ester
InChI Key XDLMVUHYZWKMMD-UHFFFAOYSA-N
SMILES CC(=C)C(=O)OCCC[Si](OC)(OC)OC
IUPAC Name 3-trimethoxysilylpropyl 2-methylprop-2-enoate
Molecular Weight (g/mol) 248.35
PubChem CID 17318
Formula Weight 248.35g/mol
Physical Form Liquid