accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 253120-47-7

IUPAC Name: 2-amino-3-(methoxycarbonyl)benzoic acid
Synonyms: 2-amino-3-(methoxycarbonyl)benzoic acid
Molecular Weight (g/mol): 195.17
Molecular Formula: C9H9NO4
InChi Key: UXQCFHRNRAGKOY-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=CC(C(O)=O)=C1N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results