accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ONH2

CAS RN 25519-80-6

IUPAC Name: 4-benzoylpiperidin-1-ium
Synonyms: 4-benzoylpiperidin-1-ium
Molecular Weight (g/mol): 190.27
Molecular Formula: C12H16NO
InChi Key: DDRCHUGHUHZNKZ-UHFFFAOYSA-O
SMILES: O=C(C1CC[NH2+]CC1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (3)
  • (2)
Quantity 
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)