accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 25519-82-8

H3COOHNHCl

CAS RN 25519-82-8

IUPAC Name: 4-(4-methoxybenzoyl)piperidine hydrochloride
Synonyms: 4-(4-methoxybenzoyl)piperidine hydrochloride
Molecular Weight (g/mol): 255.74
Molecular Formula: C13H18ClNO2
InChi Key: BBDTWYQCXXFKDH-UHFFFAOYSA-N
SMILES: Cl.COC1=CC=C(C=C1)C(=O)C1CCNCC1
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results