accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

IICH3NH3CCH3NNH2H2N

CAS RN 25535-16-4

IUPAC Name: 3,8-diamino-5-{3-[diethyl(methyl)azaniumyl]propyl}-6-phenylphenanthridin-5-ium diiodide
Synonyms: propidium iodide propidium diiodide
Molecular Weight (g/mol): 668.41
Molecular Formula: C27H34I2N4
InChi Key: XJMOSONTPMZWPB-UHFFFAOYSA-M
SMILES: [I-].[I-].CC[N+](C)(CC)CCC[N+]1=C(C2=CC=CC=C2)C2=CC(N)=CC=C2C2=CC=C(N)C=C12
Quantity 
  • (1)
  • (2)
  • (1)
  • (3)
  • (3)
  • (4)
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (11)
  • (5)
Percent Purity 
  • (8)
  • (2)
Physical Form 
  • (2)
  • (1)
  • (1)
Color 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (1)
  • (3)
  • (2)
  • (1)
  • (3)
  • (3)
  • (6)
  • (1)
  • (2)
  • (1)
  • (3)
  • (3)
  • (4)
  • (1)
  • (2)
  • (11)
  • (5)
  • (8)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)