Learn More
STEMCELL Technologies Propidium Iodide
Product Code. 17180352
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17180352 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17180352 Supplier STEMCELL Technologies Supplier No. 75002
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Cell viability dye (DNA-labeling dye)
  • Propidium Iodide (PI) is a red-fluorescent cell viability dye which is excluded from live cells with intact membranes, but penetrates dead or damaged cells and binds to DNA and RNA by intercalating between the bases
  • It is widely used as a counterstain to differentiate and exclude non-viable cells in flow cytometric analyses, and can be excited using blue (488 nm), green (532 nm), or yellow-green (561 nm) laser lines, with detection in the FL2 or FL3 channels
  • PI is used in DNA fluorescence imaging applications to discriminate early and late stages of apoptosis, to study cell‐mediated cytotoxicity, and for chromosome analysis
  • It is also commonly used in quantitative DNA assays
Specifications
CAS 25535-16-4
Quantity 10 mg