accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 2566-44-1

OH

CAS RN 2566-44-1

IUPAC Name: 2-cyclopropylethan-1-ol
Synonyms: 2-cyclopropylethanol
Molecular Weight (g/mol): 86.13
Molecular Formula: C5H10O
InChi Key: LUNMJRJMSXZSLC-UHFFFAOYSA-N
SMILES: OCCC1CC1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)