accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NOOONOOONOOOAl

CAS RN 25838-59-9

IUPAC Name: aluminium(3+) hydrate trinitrate
Synonyms: aluminium(3+) hydrate trinitrate
Molecular Weight (g/mol): 231.01
Molecular Formula: AlH2N3O10
InChi Key: ZZCONUBOESKGOK-UHFFFAOYSA-N
SMILES: O.[Al+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
Physical Form 
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)