accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 258518-65-9

NONH2N

CAS RN 258518-65-9

IUPAC Name: 3-amino-1,2-oxazole-4-carbonitrile
Synonyms: 3-amino-1,2-oxazole-4-carbonitrile
Molecular Weight (g/mol): 109.09
Molecular Formula: C4H3N3O
InChi Key: LNFYBZNMQYBIIN-UHFFFAOYSA-N
SMILES: NC1=NOC=C1C#N
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)