accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

FFSClOO

CAS RN 26120-86-5

IUPAC Name: 2,5-difluorobenzene-1-sulfonyl chloride
Synonyms: 2,5-difluorobenzenesulfonyl chloride
Molecular Weight (g/mol): 212.59
Molecular Formula: C6H3ClF2O2S
InChi Key: CELLJWUVMKEJDY-UHFFFAOYSA-N
SMILES: FC1=CC=C(F)C(=C1)S(Cl)(=O)=O
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)