missing translation for 'onlineSavingsMsg'
Learn More

2,5-Difluorobenzenesulfonyl chloride, 98%, Thermo Scientific™

Product Code. 11339915
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11339915 5 g 5g
11329915 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11339915 Supplier Thermo Scientific Alfa Aesar Supplier No. L19890.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26120-86-5
Molecular Formula C6H3ClF2O2S
Molecular Weight (g/mol) 212.595
MDL Number MFCD00060682
InChI Key CELLJWUVMKEJDY-UHFFFAOYSA-N
Synonym 2,5-difluorobenzene-1-sulfonyl chloride, 2,5-difluorobenzenesulphonyl chloride, benzenesulfonyl chloride, 2,5-difluoro, 2,5-difluoro-benzenesulfonyl chloride, 2,5-difluorobenzenesulphonylchloride, pubchem5123, acmc-1ch2j, ksc201g8t, zerenex zx009573
PubChem CID 2736911
IUPAC Name 2,5-difluorobenzenesulfonyl chloride
SMILES C1=CC(=C(C=C1F)S(=O)(=O)Cl)F

Specifications

Density 1.58
Boiling Point 221°C
Flash Point >110°C (230°F)
Refractive Index 1.516
Quantity 5 g
UN Number UN3265
Beilstein 2723886
Sensitivity Moisture sensitive
Formula Weight 212.6
Percent Purity 98%
Chemical Name or Material 2,5-Difluorobenzenesulfonyl chloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.