accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OFFFF

CAS RN 261951-79-5

IUPAC Name: 1-fluoro-3-methoxy-5-(trifluoromethyl)benzene
Synonyms: 1-fluoro-3-methoxy-5-(trifluoromethyl)benzene
Molecular Weight (g/mol): 194.13
Molecular Formula: C8H6F4O
InChi Key: OTIIFKBTTATHNT-UHFFFAOYSA-N
SMILES: COC1=CC(F)=CC(=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results