Learn More
3-Fluoro-5-(trifluoromethyl)anisole, 97%, Thermo Scientific™
Product Code. 11326699
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11326699 5 g 5g
11316699 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11326699 Supplier Thermo Scientific Alfa Aesar Supplier No. H26240.06
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 261951-79-5
Molecular Formula C8H6F4O
Molecular Weight (g/mol) 194.129
MDL Number MFCD01631491
InChI Key OTIIFKBTTATHNT-UHFFFAOYSA-N
Synonym 3-fluoro-5-trifluoromethyl anisole, 1-fluoro-3-methoxy-5-trifluoromethyl benzene, pubchem8289, acmc-1ce2b, 3-fluoro-5-methoxybenzotrifluoride, 3-fluoro-5-methoxy-1-trifluoromethyl benzene, 3-fluoro-5-trifluoromethyl phenyl methyl ether, 1-fluoranyl-3-methoxy-5-trifluoromethyl benzene, benzene,1-fluoro-3-methoxy-5-trifluoromethyl
PubChem CID 2774773
IUPAC Name 1-fluoro-3-methoxy-5-(trifluoromethyl)benzene
SMILES COC1=CC(=CC(=C1)C(F)(F)F)F
Specifications
Flash Point 52°C (126°F)
Refractive Index 1.43
Quantity 5 g
UN Number UN1993
Formula Weight 194.13
Percent Purity 97%
Chemical Name or Material 3-Fluoro-5-(trifluoromethyl)anisole