accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 2623-33-8

CH3ONHOCH3O

CAS RN 2623-33-8

IUPAC Name: 4-acetamidophenyl acetate
Synonyms: P-acetoxyacetanilide
Molecular Weight (g/mol): 193.2
Molecular Formula: C10H11NO3
InChi Key: UJAOSPFULOFZRR-UHFFFAOYSA-N
SMILES: CC(=O)NC1=CC=C(OC(C)=O)C=C1
Certifications/Compliance 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
Industry Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results