missing translation for 'onlineSavingsMsg'
Learn More

4-Acetamidophenyl acetate, 98%, BLD Pharm™

Product Code. 30943157
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
100 g
Unit Size:
100g
10g
1g
25g
5g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30943157 10 g 10g
30941103 1 g 1g
30942251 25 g 25g
30933416 100 g 100g
30936540 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 30943157 Supplier BLD Pharm Supplier No. BD14238010g
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 2623-33-8
Molecular Formula C10H11NO3
MDL Number MFCD00059205
Quantity 10 g
SMILES CC(NC1=CC=C(OC(C)=O)C=C1)=O
Molecular Weight (g/mol) 193.20
Percent Purity 98%
Chemical Name or Material 4-Acetamidophenyl acetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.