accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 2623-91-8

CH3(R)NH2OHO

CAS RN 2623-91-8

IUPAC Name: (2R)-2-aminobutanoic acid
Synonyms: D-2-aminobutyric acid D-2-aminobuttersaeure delta-2-aminobutanoic acid delta-2-aminobuttersaeure delta-(-)-2-aminobutyric acid delta-2-aminobutyrate delta-2-aminobutyric acid
Molecular Weight (g/mol): 103.12
Molecular Formula: C4H9NO2
InChi Key: QWCKQJZIFLGMSD-GSVOUGTGSA-N
SMILES: CC[C@@H](N)C(O)=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)