missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(R)-2-Aminobutyric Acid, TRC
Click to view available options
Quantity:
5 g
Unit Size:
5g
Specifications
Specifications
| Chemical Name or Material | (R)-2-Aminobutyric Acid |
| CAS | 2623-91-8 |
| Molecular Formula | C4H9NO2 |
| Quantity | 5 g |
| Synonym | (R)-2-Amino-butanoic Acid, D-2-Amino-butyric Acid, (-)-2-Aminobutanoic Acid, (-)-2-Aminobutyric Acid, (-)-alpha-Aminobutyric Acid, (2R)-2-Aminobutanoic Acid, (2R)-2-Aminobutyric Acid, (R)-(-)-alpha-Aminobutyric Acid, (R)-2-Aminobutanoic Acid, (R)-alpha-Aminobutyric Acid, D-2-Amino-n-butyric Acid, D-2-Aminobutanoic Acid, D-2-Aminobutyric Acid, D-Butyrine, D-Ethylglycine, D-alpha-Amino-n-butyric Acid, D-alpha-Aminobutyric Acid |
| InChI Formula | InChI=1S/C4H9NO2/c1-2-3(5)4(6)7/h3H,2,5H2,1H3,(H,6,7)/t3-/m1/s1 |
| SMILES | CC[C@@H](N)C(=O)O |
| IUPAC Name | (2R)-2-aminobutanoic acid |
| Molecular Weight (g/mol) | 103.12 |
| Formula Weight | 103.0633 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction