accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3CH3NHO(S)CH3HNOOHCH3CH3NHO(S)HNO(S)NHOH3CCH3H3CCH3H3CCH3OHOHO

CAS RN 26305-03-3

IUPAC Name: 3-hydroxy-4-[(2S)-2-{3-hydroxy-6-methyl-4-[(2S)-3-methyl-2-[(2S)-3-methyl-2-(3-methylbutanamido)butanamido]butanamido]heptanamido}propanamido]-6-methylheptanoic acid
Synonyms: pepstatin
Molecular Weight (g/mol): 685.9
Molecular Formula: C34H63N5O9
InChi Key: FAXGPCHRFPCXOO-JHTFMCFMSA-N
SMILES: CC(C)CC(NC(=O)[C@H](C)NC(=O)CC(O)C(CC(C)C)NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)CC(C)C)C(C)C)C(C)C)C(O)CC(O)=O
Physical Form 
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Form 
  • (2)
Product Type 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (2)
  • (2)
  • (1)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)