Learn More
Pepstatin A, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 26305-03-3 |
| Molecular Formula | C34H63N5O9 |
| MDL Number | MFCD00060740 |
| Quantity | 250 mg |
| SMILES | CC(C)CC(N[C@@H](C(C)C)C(N[C@@H](C(C)C)C(N[C@@H](CC(C)C)[C@@H](O)CC(N[C@@H](C)C(N[C@@H](CC(C)C)[C@@H](O)CC(O)=O)=O)=O)=O)=O)=O |
| Molecular Weight (g/mol) | 685.89 |
| Percent Purity | 98% |
| Chemical Name or Material | Pepstatin A |