accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3H3COF

CAS RN 26393-91-9

IUPAC Name: 1-(4-fluorophenyl)-2-methylpropan-1-one
Synonyms: 1-(4-fluorophenyl)-2-methylpropan-1-one
Molecular Weight (g/mol): 166.2
Molecular Formula: C10H11FO
InChi Key: MHUVRVXSYXJUPK-UHFFFAOYSA-N
SMILES: CC(C)C(=O)C1=CC=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results