accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3NNCH3OCl

CAS RN 27006-76-4

IUPAC Name: 5-chloro-1,3-dimethyl-1H-pyrazole-4-carbaldehyde
Synonyms: 5-chloro-1,3-dimethylpyrazole-4-carbaldehyde
Molecular Weight (g/mol): 158.59
Molecular Formula: C6H7ClN2O
InChi Key: SZRSMNYUEXXEBL-UHFFFAOYSA-N
SMILES: CN1N=C(C)C(C=O)=C1Cl
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)