accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

(R)(R)HNNHNHHNOOOO(S)(S)OOOOSSOHOHNH2NH2HOHO

CAS RN 27025-41-8

IUPAC Name: 2-amino-4-[(2-{[2-(4-amino-4-carboxybutanamido)-2-[(carboxymethyl)carbamoyl]ethyl]disulfanyl}-1-[(carboxymethyl)carbamoyl]ethyl)carbamoyl]butanoic acid
Synonyms: oxidized glutathione oxiglutatione 奥谷胱甘肽 oxidised glutathione
Molecular Weight (g/mol): 612.63
Molecular Formula: C20H32N6O12S2
InChi Key: YPZRWBKMTBYPTK-UHFFFAOYNA-N
SMILES: NC(CCC(=O)NC(CSSCC(NC(=O)CCC(N)C(O)=O)C(=O)NCC(O)=O)C(=O)NCC(O)=O)C(O)=O
Quantity 
  • (4)
  • (1)
  • (1)
  • (5)
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (3)
  • (2)
  • (3)
  • (2)
Physical Form 
  • (3)
  • (3)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (3)
  • (2)
  • (4)
  • (1)
  • (1)
  • (5)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (3)
  • (4)