missing translation for 'onlineSavingsMsg'
Learn More

Glutathione, ≥95%, Oxidized, Thermo Scientific™

Product Code. 15805158 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Plastic bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15805158 1 g Plastic bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15805158 Supplier Thermo Scientific Supplier No. J16320.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 27025-41-8
Packaging Plastic bottle
Molecular Formula C20H32N6O12S2
MDL Number MFCD00150701
Quantity 1 g
Beilstein 1718700
Synonym oxiglutatione, glutathione disulfide, gssg, oxidized glutathione, glutathione disulphide, glutathione-ssg, l-glutathione oxidized, oxigluthione, glutathone disulfide, glutathione oxidized
InChI Key YPZRWBKMTBYPTK-IOKZUGQQNA-N
SMILES N[C@@H](CCC(=O)N[C@@H](CSSC[C@H](NC(=O)CC[C@H](N)C(O)=O)C(=O)NCC(O)=O)C(=O)NCC(O)=O)C(O)=O
IUPAC Name 2-amino-4-[(2-{[2-(4-amino-4-carboxybutanamido)-2-[(carboxymethyl)carbamoyl]ethyl]disulfanyl}-1-[(carboxymethyl)carbamoyl]ethyl)carbamoyl]butanoic acid
Molecular Weight (g/mol) 612.63
PubChem CID 65359
ChEBI CHEBI:17858
Formula Weight 612.63
Percent Purity ≥95%
Physical Form Lyophilized Powder
Chemical Name or Material Glutathione
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.