accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3ONHOHO

CAS RN 27115-49-7

IUPAC Name: 2-[(3-methylphenyl)formamido]acetic acid
Synonyms: [(3-methylphenyl)formamido]acetic acid
Molecular Weight (g/mol): 193.2
Molecular Formula: C10H11NO3
InChi Key: YKAKNMHEIJUKEX-UHFFFAOYSA-N
SMILES: CC1=CC=CC(=C1)C(=O)NCC(O)=O
Molecular Weight (g/mol) 
  • (3)
  • (4)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (3)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (3)
  • (4)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)