missing translation for 'onlineSavingsMsg'
Learn More

3-Methylhippuric acid, 98+%, ACROS Organics™

Product Code. 10195150 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10195150 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10195150 Supplier Thermo Scientific Acros Supplier No. 259890010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 27115-49-7
CAS Min % 98.0
CAS Max % 100.0
Melting Point 138°C to 142°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass bottle
Molecular Formula C10H11NO3
Linear Formula CH3C6H4CONHCH2COOH
MDL Number MFCD00044399
Quantity 1 g
Synonym 3-methylhippuric acid, m-methylhippuric acid, n-m-toluoyl glycine, m-toluric acid, n-3-methylbenzoyl glycine, 3-methylbenzoylglycine, glycine, n-3-methylbenzoyl, meta-methylhippuric acid, hippuric acid, m-methyl, 3-methyl hippuric acid
InChI Key YKAKNMHEIJUKEX-UHFFFAOYSA-N
SMILES CC1=CC=CC(=C1)C(=O)NCC(O)=O
IUPAC Name 2-[(3-methylphenyl)formamido]acetic acid
Molecular Weight (g/mol) 193.20
PubChem CID 99223
ChEBI CHEBI:68500
Formula Weight 193.19
Percent Purity ≥98.0%
Physical Form Crystalline Powder
Chemical Name or Material 3-Methylhippuric acid, 98+%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.