accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 27176-10-9

H2OOPOOOKKK

CAS RN 27176-10-9

IUPAC Name: tripotassium hydrate phosphate
Synonyms: tripotassium hydrate phosphate
Molecular Weight (g/mol): 230.28
Molecular Formula: H2K3O5P
InChi Key: RMNIZOOYFMNEJJ-UHFFFAOYSA-K
SMILES: O.[K+].[K+].[K+].[O-]P([O-])([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
Physical Form 
  • (2)
  • (5)
  • (1)
Grade 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (5)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (5)
  • (1)
  • (5)