accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 27208-37-3

CAS RN 27208-37-3

IUPAC Name: pentacyclo[12.3.1.0⁴,¹⁷.0⁷,¹⁶.0¹⁰,¹⁵]octadeca-1(18),2,4(17),5,7,9,11,13,15-nonaene
Synonyms: pentacyclo[12.3.1.0⁴,¹⁷.0⁷,¹⁶.0¹⁰,¹⁵]octadeca-1(18),2,4(17),5,7,9,11,13,15-nonaene
Molecular Weight (g/mol): 226.28
Molecular Formula: C18H10
InChi Key: BZCXQYVNASLLQO-UHFFFAOYSA-N
SMILES: C1=CC2=C3C1=CC1=CC=CC4=CC=C(C=C2)C3=C14
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)