missing translation for 'onlineSavingsMsg'
Learn More

Acepyrene, TRC

Product Code. 17943445
Change view
Click to view available options
Quantity:
0.5 mg
1 mg
2.5 mg
Unit Size:
0.5mg
1mg
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17943445 1 mg 1mg
17921035 2.5 mg 2.5mg
17903325 0.5 mg 0.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17943445 Supplier Toronto Research Chemicals Supplier No. TRCA1309501MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27208-37-3
Molecular Formula C18 H10
Molecular Weight (g/mol) 226.27
Synonym Cyclopenta[cd]pyrene, Acepyrene, Acepyrylene, Cyclopenta[c,d]pyrene
IUPAC Name pentacyclo[12.3.1.04,17.07,16.010,15]octadeca-1(18),2,4(17),5,7(16),8,10(15),11,13-nonaene
SMILES C1=Cc2cc3cccc4ccc5ccc1c2c5c34

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C18H10/c1-2-11-4-6-13-7-5-12-8-9-15-10-14(3-1)16(11)18(13)17(12)15/h1-10H
Formula Weight 226.0783
Percent Purity >95%
Chemical Name or Material Cyclopenta(c,d)pyrene
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.